Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Closure Size | Description | Liner Material | No. per Case |
---|---|---|---|---|
S297-5300 | 53-400 mm | Open top septum cap | Bonded 0.045 in. PTFE-lined silicone septa | 24 |
297-2400UB | 24-400 mm | Open top septum cap | Bonded 0.060 in. thin PTFE-lined silicone septa | 24 |
297-2414 | 24-414 mm | Open top cap | None | 24 |
24-414WS-3M also known as 24-414/WS-3M | 24-414 mm | Open top septum cap | Bonded 0.125 in. PTFE-lined silicone septa | 1000 |
Thermo Scientific™ Septa Caps include a bonded-in 0.045 in. thick septa. The same high quality septa caps used on our Thermo Scientific VOA Vials, septa bottles and septa jars. Processed to meet EPA performance–based specifications for Volatile Organic Analysis.
Processed to meet EPA Performance Based Specifications for Volatile Organic Analysis
Includes:
Cap only
Compatible with:
For use with Thermo Scientific VOA Vials, bottles and jars. 297-2414 closures are compatible with loose septa Cat. Nos. 288-0022, 288-0072, 288-0022BP and S24-400-S2.
Share catalog number, name or link