Invitrogen™

SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 750

Catalog number: S30046
Have Questions?
Invitrogen™

SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 750

Catalog number: S30046
Have Questions?
Catalog Number
S30046
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S300461 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Alexa Fluor™ 750 SAIVI™ Antibody Labeling Kit provides a convenient means to label antibodies with an optimal degree of labeling for in vivo imaging applications (DOL; ∼2) over a 6-fold antibody concentration range with no adjustments in reaction volume, dye concentration, or antibody concentration necessary. Using this procedure, optimally labeled antibodies are ready for applications that require azide-free reagents, such as live-cell imaging or direct injection into animals.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Type
Labeling Kit
Detection Method
Fluorescence
Excitation/Emission
749/775
Labeling Scale
0.5 to 3 mg
Shipping Condition
Room Temperature
Product Line
Alexa Fluor™, SAIVI™
Label or Dye
Alexa Fluor 750
Color
Infrared
Labeling Target
Antibodies
Label Type
Alexa Fluor Dyes

Contents & Storage

Store in refrigerator 2°C to 8°C and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text