Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

SYBR™ Safe DNA Gel Stain in 0.5X TBE

Catalog number: S33101
Have Questions?
Invitrogen™

SYBR™ Safe DNA Gel Stain in 0.5X TBE

Catalog number: S33101
Have Questions?
Catalog Number
S33101
Unit Size
4 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S331014 L
-
S331001 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SYBR Safe DNA gel stain was developed specifically for reduced mutagenicity – to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels. SYBR Safe stain is not only less mutagenic than ethidium bromide, but SYBR Safe stain's detection sensitivity is better than that of ethidium bromide. SYBR Safe stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe stain has fluorescence excitation maxima at ∼280 and ∼502 nm, and an emission maximum at ∼530 nm. SYBR Safe DNA gel stain is also available as 1 L (S-33100) of ready-to-use solution. The SYBR Safe DNA Gel Stain Starter Kit (S-33110) includes 1 L of SYBR Safe DNA gel stain and one photographic filter (S-37100).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescence
Label or Dye
SYBR Safe
Product Type
DNA Gel Stain
Shelf Life
6 months from date of purchase
Green Features
Less hazardous
Quantity
4 L
Shipping Condition
Room Temperature
Target Molecule
DNA

Contents & Storage

• Supplied as a ready-to-use solution in 0.5X TBE

Store at room temperature in the original container.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text