Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

SYBR™ Safe DNA Gel Stain in 1X TAE

Catalog number: S33112
Have Questions?
Invitrogen™

SYBR™ Safe DNA Gel Stain in 1X TAE

Catalog number: S33112
Have Questions?
Catalog Number
S33112
Unit Size
4 L
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 L
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S331124 L
-
S331111 L
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
SYBR Safe™ DNA gel stain was developed specifically for reduced mutagenicity—to be safer than ethidium bromide for staining DNA in agarose or acrylamide gels. SYBR Safe™stain is not only less mutagenic than ethidium bromide, but SYBR Safe™ stain's detection sensitivity is better than that of ethidium bromide. SYBR Safe™ stain comes as a premixed solution that can be used just like an ethidium bromide solution, either in the gel during the electrophoresis run or as a post-run stain. Bound to nucleic acids, the green-fluorescent SYBR Safe™ stain has fluorescence excitation maxima at ∼280 and ∼502 nm, and an emission maximum at ∼530 nm.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescence
Label or Dye
SYBR Safe
Product Type
DNA Gel Stain
Shelf Life
6 months from date of purchase
Green Features
Less hazardous
Quantity
4 L
Shipping Condition
Room Temperature
Target Molecule
DNA

Contents & Storage

• Supplied as a ready-to-use solution in 1X TAE

Store at room temperature in the original container.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text