1x1 image pixel for data collection
Singlet Oxygen Sensor Green - Special Packaging
Singlet Oxygen Sensor Green - Special Packaging
Invitrogen™

Singlet Oxygen Sensor Green - Special Packaging

Singlet Oxygen Sensor Green is a detection reagent that is highly selective for singlet oxygen. Unlike other available fluorescent andRead more
Have Questions?
Catalog NumberQuantity
S3600210 x 100 μg
Catalog number S36002
Price (USD)
345.65
Online Exclusive
358.00
Save 12.35 (3%)
Each
In stock
Add to cart
Quantity:
10 x 100 μg
Recurring order eligible. Learn more »
Price (USD)
345.65
Online Exclusive
358.00
Save 12.35 (3%)
Each
Add to cart
Ask our AI about this Product
Singlet Oxygen Sensor Green is a detection reagent that is highly selective for singlet oxygen. Unlike other available fluorescent and chemiluminescent singlet oxygen detection reagents, Singlet Oxygen Sensor Green does not show any appreciable response to hydroxyl radical or superoxide. This indicator initially exhibits weak blue fluorescence, but in the presence of singlet oxygen, it emits a green fluorescence (excitation/emission maxima ∼504/525 nm) similar to that of fluorescein. Singlet Oxygen Sensor Green is cell-impermeant.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Quantity10 x 100 μg
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2935447Certificate of AnalysisJul 29, 2024S36002
2761161Certificate of AnalysisNov 07, 2023S36002
2738382Certificate of AnalysisSep 20, 2023S36002
2552720Certificate of AnalysisDec 16, 2022S36002
2456925Certificate of AnalysisMar 08, 2022S36002
5 results displayed, search above for a specific certificate

Safety Data Sheets

Citations & References (23)

Citations & References
Abstract
Imaging the production of singlet oxygen in vivo using a new fluorescent sensor, Singlet Oxygen Sensor Green.
Authors:Flors C, Fryer MJ, Waring J, Reeder B, Bechtold U, Mullineaux PM, Nonell S, Wilson MT, Baker NR
Journal:J Exp Bot
PubMed ID:16595576
'Singlet oxygen is known to be produced by cells in response to photo-oxidative stresses and wounding. Due to singlet oxygen being highly reactive, it is thought to have a very short half-life in biological systems and, consequently, it is difficult to detect. A new commercially available reagent (singlet oxygen sensor ... More
Mechanisms of bacteriophage inactivation via singlet oxygen generation in UV illuminated fullerol suspensions.
Authors:Hotze EM, Badireddy AR, Chellam S, Wiesner MR,
Journal:Environ Sci Technol
PubMed ID:19764229
'Nonenveloped viruses are shown to be inactivated by singlet oxygen ((1)O2) produced in UVA photosensitized aqueous suspensions of a polyhydroxylated fullerene (C60(OH)22-24; fullerol, 40 microM). Experiments were performed with MS2, a ssRNA bacteriophage, as well as two dsDNA phages: PRD1, which has an internal lipid membrane, and T7, which entirely ... More
Monitoring singlet oxygen and hydroxyl radical formation with fluorescent probes during photodynamic therapy.
Authors:Price M, Reiners JJ, Santiago AM, Kessel D,
Journal:Photochem Photobiol
PubMed ID:19508643
'Singlet oxygen (1O2) is the primary oxidant generated in photodynamic therapy (PDT) protocols involving sensitizers resulting in type II reactions. 1O2 can give rise to additional reactive oxygen species (ROS) such as the hydroxyl radical (*OH). The current study was designed to assess 3''-p-(aminophenyl) fluorescein (APF) and 3''-p-(hydroxyphenyl) fluorescein (HPF) ... More
Singlet oxygen photosensitisation by the fluorescent probe Singlet Oxygen Sensor Green.
Authors:Ragàs X, Jiménez-Banzo A, Sánchez-García D, Batllori X, Nonell S,
Journal:Chem Commun (Camb)
PubMed ID:19436910
The fluorescent probe Singlet Oxygen Sensor Green is able to produce singlet oxygen under exposure to UV or visible radiation.
Imaging of reactive oxygen species in vivo.
Authors:Driever SM, Fryer MJ, Mullineaux PM, Baker NR,
Journal:Methods Mol Biol
PubMed ID:19083172
Reactive oxygen species (ROS) are involved in many signalling pathways and numerous stress responses in plants. Consequently, it is important to be able to identify and localize ROS in vivo to evaluate their roles in signalling. A number of probes that have a high affinity for specific ROS and that ... More
23 total citations

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection