SlowFade™ Gold Antifade Mountant with DAPI
SlowFade™ Gold Antifade Mountant with DAPI
Invitrogen™

SlowFade™ Gold Antifade Mountant with DAPI

Green features
SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
S3693810 mL
S369395 x 2 mL
S369422 mL
3 Options
Catalog number S36938
Price (USD)/ 10 mL
241.00
-
Add to cart
Quantity:
10 mL
Price (USD)/ 10 mL
241.00
Add to cart
SlowFade™ Gold Antifade Mountant with DAPI
Catalog numberS36938
Price (USD)/ 10 mL
241.00
-
Add to cart

SlowFade Gold Antifade Mountant is a liquid mountant applied directly to fluorescently labeled cell or tissue samples on microscope slides. It contains chemical components designed to protect fluorescent dyes from fading (photobleaching) during fluorescence microscopy experiments. SlowFade Gold Antifade Mountant is glycerol-based, with no mixing or curing required, which makes it ideal for immediate viewing of the sample. It comes ready-to-use—just apply a drop to the sample, add a coverslip, and image. It is available with or without DAPI nuclear stain.

Key attributes:
• Protects dyes from fading during imaging
• Ready-to-use liquid, ideal for immediate sample viewing
• Mounted samples are stable for weeks
• Maintains signal strength—little to no quenching
• Ideal for Alexa Fluor dyes

Select the antifade mounting or optical clearing reagent that matches your fluorescence imaging needs ›

SlowFade Gold Antifade Mountant is not recommended for mounting samples containing fluorescent proteins, such as GFP. For superior antifade protection of fluorescent proteins and fluorescent dyes in cultured cells or thin tissue slices, SlowFade Diamond Antifade Mountant is recommended. For high resolution or to image thicker tissue or 3D cell culture with a focal depth of 0–500 μm, try SlowFade Glass Antifade Mountant.

Select best SlowFade antifade reagent for your experimental needs ›

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Microscope
Product LineSlowFade™
Product TypeMounting Media
Quantity10 mL
Solution TypeAnti-fade Mountant
Green FeaturesLess hazardous, sustainable packaging
Label or DyeDAPI
Shipping ConditionRoom Temperature
FormLiquid
Unit Size10 mL
Contents & Storage
Storage at room temperature is recommended but can also be stored frozen (-5° to -30°C). Protect from light.