Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Shelf Life | 1 Year 3 Months |
Health Hazard 1 | Serious eye damage/eye irritation (category 1), Corrosive to metals (category 1), Skin corrosion/irritation (category 2) |
Quantity | 500 mL |
Health Hazard 2 | Causes serious eye damage, May be corrosive to metals, Causes skin irritation |
UN Number | UN3264 |
Physical Form | Liquid |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
S55602.AP also known as S55602-AP | Each | 500 mL | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|