iCAP TQ Tune Solution (BRE0009578)
Product Image
Thermo Scientific™

iCAP TQ Tune Solution (BRE0009578)

 
Have Questions?
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number S55611.AP
also known as S55611-AP
Price (USD)/ Each
360.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
424.00 
Save 63.35 (15%)
-
Add to cart
Quantity:
500 mL
Price (USD)/ Each
360.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
424.00 
Save 63.35 (15%)
Add to cart
iCAP TQ Tune Solution (BRE0009578)
Catalog numberS55611.AP
Price (USD)/ Each
360.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
424.00 
Save 63.35 (15%)
-
Add to cart
 
RUO – Research Use Only
Specifications
Shelf Life1 Year 3 Months
Health Hazard 1Serious eye damage/eye irritation (category 1), Corrosive to metals (category 1), Skin corrosion/irritation (category 2)
Quantity500 mL
Health Hazard 2Causes serious eye damage, May be corrosive to metals, Causes skin irritation
UN NumberUN3264
Physical FormLiquid
Unit SizeEach