1x1 image pixel for data collection
Oxoid™ IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide
Product Image
Thermo Scientific™
Oxoid™ IMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide
Oxoid IMAGEN Respiratory Syncytial Virus Positive Control is a slide with acetone-fixed human epithelial cells infected with RSV.
Have Questions?
Catalog NumberQuantity
S610830-21 Slide/Pk.
Catalog number S610830-2
Price (USD)
-
Request A Quote
Quantity:
1 Slide/Pk.
The IMAGEN™ Control Slides are for use as a control to the following tests: IMAGEN Adenovirus (K610011-2) IMAGEN Respiratory Syncyti al Virus (RSV) (K610211-2) IMAGEN Parainfluenza Group (K610311-2) IMAGEN Parainfluenza Typing 1, 2 and 3 (K610411-2) IMAGEN Influenza A and B Virus (K610511-2) IMAGEN Respiratory Screen (K612011-2) IMAGEN hMPV (K612511-2) IMAGEN Herpes Simplex Virus (HSV) (K610611-2) IMAGEN Chlamydia kit (K610111-2)
Sealed in foil pouch filled with nitrogen.
Specifications
DescriptionIMAGEN™ Respiratory Syncytial Virus (RSV) Positive Control Slide
Quantity1 Slide/Pk.
TypeControl
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link