Informational:Inquire about OEM or Commercial Supply version of this product here.
Invitrogen™

SYBR™ Green Nucleic Acid Gel Stain Starter Kit

Catalog number: S7580
Have Questions?
Invitrogen™

SYBR™ Green Nucleic Acid Gel Stain Starter Kit

Catalog number: S7580
Have Questions?
Catalog Number
S7580
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
S75801 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The SYBR Green Nucleic Acid Stain Starter Kit makes it easy for first-time users to try our two SYBR Green gel stains. The kit contains: 50 μL of SYBR Green I nucleic acid gel stain (S-7567), 50 μL of SYBR Green II RNA gel stain (S-7568) and a SYBR Green/Gold gel stain photographic filter (S-7569).
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Location
In-Gel Detection
Detection Method
Fluorescence
Label or Dye
SYBR Green II, SYBR Green I
Product Type
Nucleic Acid Gel Stain Starter Kit
Target Molecule
RNA, DNA

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text