SYBR™ Green I Nucleic Acid Gel Stain is one of the most sensitive stains available for detecting double-stranded DNA (dsDNA) in agarose and polyacrylamide gels. Because SYBR™ Green I has greater sensitivity for dsDNA, it is especially useful for assays where the presence of contaminating RNA or ssDNA might obscure results. With exceptionally low background fluorescence and spectral characteristics that closely match light sources and filter sets in existing instruments, SYBR™ Green I stain is ideal for use with laser scanners. SYBR™ Green I stain provides:
• Sensitivity—at least four times greater than ethidium bromide for DNA in agarose gels • Ease of use—gels soaked in diluted stain can be visualized without desalting • Compatibility—with UV transilluminators, gel documentation systems, and laser scanners • Flexibility—for use with a broad range of applications, including DNA typing, PCR-based assays, DNA damage assays, analysis of complex samples, and real-time PCR detection
Product use One mL stains 100 minigels. Various pack sizes are available.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Location
In-Gel Detection
Detection Method
Fluorescence
Label or Dye
SYBR Green I
Product Type
Nucleic Acid Gel Stain
Quantity
20 x 50 μL
Shipping Condition
Room Temperature
Target Molecule
DNA
Contents & Storage
• Supplied as a 10,000X concentrate in DMSO
Store at -20°C, protected from light in a dessicator.