Streptavidin, solid
Streptavidin, solid
Invitrogen™

Streptavidin, solid

Streptavidin is a bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.Streptavidin is unlabeled, unconjugated, and providedRead more
Have Questions?
Catalog NumberQuantity
S8885 mg
Catalog number S888
Price (TWD)
14,270.00
Online offer
Ends: 30-Jun-2025
16,700.00
Save 2,430.00 (15%)
Each
-
Add to cart
Quantity:
5 mg
Recurring order eligible. Learn more »
Price (TWD)
14,270.00
Online offer
Ends: 30-Jun-2025
16,700.00
Save 2,430.00 (15%)
Each
Add to cart
Ask our AI about this Product
Streptavidin is a bacterially derived biotin-binding protein that reportedly exhibits less nonspecific binding than avidin.

Streptavidin is unlabeled, unconjugated, and provided as a lyophilized powder.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeStreptavidin Conjugate (fluorescent)
Quantity5 mg
Shipping ConditionRoom Temperature
ConjugateUnconjugated
FormSolid
Unit SizeEach
Contents & Storage
Storage Upon Receipt:≤-20°C, desiccate
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Kunden, die diesen Artikel ansahen, interessierten sich auch für



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3192379Certificate of AnalysisJun 20, 2025S888
2920645Certificate of AnalysisJul 11, 2024S888
2551350Certificate of AnalysisJan 25, 2023S888
2418504Certificate of AnalysisNov 03, 2021S888
2306804Certificate of AnalysisMar 31, 2021S888
5 results displayed, search above for a specific certificate

Safety Data Sheets

Frequently asked questions (FAQs)

Streptavidin is a protein composed of four identical subunits, each containing a high affinity binding site for biotin (K-D = 10 -15 M) . Streptavidin has the same biotin binding properties as avidin, but it has a low isoelectric point (pI=5) and no carbohydrate groups, resulting in low non-specific binding.

Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.

Although we utilize manufacturing methods that minimize the introduction of any contaminants such as RNases, we do not process this product to specifically remove or inactivate RNases and do not test for RNase activity.

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.

Citations & References (18)

Citations & References
Abstract
Easily reversible desthiobiotin binding to streptavidin, avidin, and other biotin-binding proteins: uses for protein labeling, detection, and isolation.
Authors:Hirsch JD, Eslamizar L, Filanoski BJ, Malekzadeh N, Haugland RP, Beechem JM, Haugland RP
Journal:Anal Biochem
PubMed ID:12419349
'The high-affinity binding of biotin to avidin, streptavidin, and related proteins has been exploited for decades. However, a disadvantage of the biotin/biotin-binding protein interaction is that it is essentially irreversible under physiological conditions. Desthiobiotin is a biotin analogue that binds less tightly to biotin-binding proteins and is easily displaced by ... More
Immunochemical analysis shows all three domains of diphtheria toxin penetrate across model membranes.
Authors:Tortorella D, Sesardic D, Dawes CS, London E
Journal:J Biol Chem
PubMed ID:7499201
'Diphtheria toxin undergoes membrane insertion and translocation across membranes when exposed to low pH. In this study, the translocation of the toxin has been investigated by the binding of antibodies to two preparations of model membrane-inserted toxin. In one preparation, toxin was added externally to model membrane vesicles and then ... More
Cell-adhesive properties of streptavidin are mediated by the exposure of an RGD-like RYD site.
Authors:Alon R, Bayer EA, Wilchek M
Journal:Eur J Cell Biol
PubMed ID:1425765
'The interaction of streptavidin with various cell systems was studied using fluorescent derivatives of the protein. The native unprocessed form of streptavidin bound to cells at low levels and in a nonspecific manner. In contrast, both the truncated "core" streptavidin (the commercially available form) and the biotin-blocked unprocessed protein bound ... More
Cell adhesion to streptavidin via RGD-dependent integrins.
Authors:Alon R, Bayer EA, Wilchek M
Journal:Eur J Cell Biol
PubMed ID:8462588
'Biotin-blocked streptavidin binds specifically (Kd approximately 3 x 10(-8) M) to cell surfaces, presumably via an RYD-containing sequence. This site is distinct from the biotin-binding cleft of the protein and bears high homology to the RGD-containing cell-binding domain of fibronectin. We show here that various cell types adhere to immobilized ... More
A fluorescence resonance energy transfer-based approach for investigating late endosome-lysosome retrograde fusion events.
Authors:Kaufmann AM, Goldman SD, Krise JP,
Journal:Anal Biochem
PubMed ID:19109922
'Traditionally, lysosomes have been considered to be a terminal endocytic compartment. Recent studies suggest that lysosomes are quite dynamic, being able to fuse with other late endocytic compartments as well as with the plasma membrane. Here we describe a quantitative fluorescence energy transfer (FRET)-based method for assessing rates of retrograde ... More
18 total citations

Andere Produkte



Share catalog number, name or link

1x1 image pixel for data collection