You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Streptavidin, (PE-Cy7)
Invitrogen™
Streptavidin, (PE-Cy7)
• Class: RUO• Statement: For research use only. Not for use in diagnostic procedures.• Buffer: PBS• Preservative: 0.1% Sodium
AzideView
Read more
Have Questions?
Catalog number
SA1012
Price (USD)
/ 0.5 mL
373.00
-
Add to cart
Price (USD)
/ 0.5 mL
373.00
Add to cart
Streptavidin, (PE-Cy7)
Catalog number
SA1012
Price (USD)
/ 0.5 mL
373.00
-
Add to cart
•
Class:
RUO
•
Statement:
For research use only. Not for use in diagnostic procedures.
•
Buffer:
PBS
•
Preservative:
0.1% Sodium Azide
View a
selection guide for all streptavidin RPE, APC, and tandem conjugates
.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration
1 mg/mL
Conjugate
PE-Cy7
Detection Method
Fluorescence
For Use With (Equipment)
Flow Cytometry, Immunoassays, Histochemical Applications, Blot Analysis
Quantity
0.5 mL
Excitation/Emission
488/767
Flow Cytometer Laser Lines
488, 488 or 561, 561
Shipping Condition
Wet Ice
Form
Solution
Product Line
Cy™
Buffer
PBS Buffer
Product Type
Streptavidin Conjugate (Fluorescent)
Label or Dye
PE-Cy7
Unit Size
0.5 mL
Contents & Storage
Contains 1 vial of streptavidin,PE-Cy7 conjugate.
Store in refrigerator (2–8°C).