Thermo Scientific™

Productainer™ BioProcess Container (BPC), 100L with 4 Top Ports, 3D Single-Use BPC

Catalog number: SH30651.02
Have Questions?
Thermo Scientific™

Productainer™ BioProcess Container (BPC), 100L with 4 Top Ports, 3D Single-Use BPC

Catalog number: SH30651.02
Have Questions?
Catalog Number
SH30651.02
Unit Size
100 L
Price (USD)
Catalog NumberUnit SizePrice (USD)
SH30651.02100 LRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific™ Productainer™ BioProcess Container (BPC), 100 L 3D system is composed of CX5-14 single web, multi-layer plastic film and includes 4 top ports. The 3D square tube design with top dispense is for use with flat-bottom Thermo Scientific™ drums.

Features:
• Use this system to eliminate post-use cleaning steps required with re-useable containers, and to reduce cross-contamination risks.
• All 3D BPCs are constructed in an ISO 7 cleanroom under cGMP conditions.
• All 3D BPCs are designed to fit the full range of support containers, both square and cylindrical – from 50 to 2000 L.

Applications of the Thermo Scientific 3D BPC include:
• Hydration and sterilization of process buffers, liquids, and culture media.
• Chromatography feed and fraction collection.
• Storage and transport of bulk drug product and bulk drug precursors.
• Harvest from and feed into bioreactors and fermentors.
• Dispensing, packaging, and storage of cell culture media, buffers and process liquids.

Customize standard BPC systems for optimized single-use technology. Choose from the industry’s largest component library – with over 20,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style.
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors.
Filters—used for bioburden reduction; sterilization grade filters and vent filters.
Tubing—thermoplastic elastomers, platinum-cured silicone.
Sample ports—septum-style and needle-free ports.
Clamps—sliders, pinch, and crimp bands.

Specifications

Description
100 L dip tube: 71 cm (28 in.)
Film Description
CX5-14
Port Detail
Top Ports: 2 x 0.25 in. 2 x 0.38 in.
For Use With (Equipment)
SV50076.03
Material
Polyethylene
Outer Packaging
Packages placed in strong cardboard cartons with product and lot identification
Protective Inner Packaging
Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Sterilization Method
Gamma Irradiated
Tubing Line 1
Line 1. MPC insert, polycarbonate, Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.) with dip tube length dependent on size
Tubing Line 2
Line 2. Insert luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Tubing Line 3
Line 3. Body luer lock connection, polypropylene Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 6.4 x 9.5 mm (0.25 x 0.38 in.)
Tubing Line 4
Line 4. MPC body, polycarbonate Tubing: C-Flex, length: 121.9 cm (48 in.) ID x OD: 9.5 x 15.9 mm (0.38 x 0.63 in.) dip tube length: 30.5 cm (12 in.)
Product Line
Productainer™
Green Features
Fewer resources used and less waste

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text