BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 3 ports
BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 3 ports
Thermo Scientific™
BioProcess Container (BPC), 2000L, for HyPerforma™ Single-Use Mixer, powder-to-liquid, Aegis™ 5-14 film, 3 ports
Green features
The 2000L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic filmRead more
Have Questions?
Catalog number SH30974.05
Price (USD)/ 2000 L
-
Request A Quote
The 2000L Thermo Scientific™ Bioprocess Containers (BPC) for powder-to-liquid mixing, is composed of Aegis 5-14 single web, multi-layer plastic film and includes 3 ports.
Single Use Mixer (S.U.M.) BPC options include:
• Open top liner for media/buffer hydration
• Closed BPC with powder port for contained media/buffer hydration
• Closed BPC with powder port and monitoring probe capability
• Closed BPC for liquid-liquid mixing of critical sterile solutions such as pooling of fractions or processing of intermediate solutions
• Closed BPC for liquid-liquid mixing with monitoring probe capability
Specifications
Green FeaturesFewer resources used and less waste
Capacity (Metric)2000 L
Port Detail3 ports
QuantityEach
Product LineHyPerforma™
Unit Size2000 L