Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, 30 L
Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, 30 L
Thermo Scientific™
Thermo Scientific™ Single-Use Fermentor BioProcess Container, CX5-14 film, pH/DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, 30 L
This 30 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin freeMás información
Have Questions?
Número de catálogoCantidad
SH310190230 L
Número de catálogo SH3101902
Precio (EUR)
-
Solicite un presupuesto
Cantidad:
30 L
Ask our AI about this Product
This 30 L Thermo Scientific Single-Use Fermentor (SUF) BioProcess Container (BPC) is composed of CX5-14 film, a five-layer, animal-origin free film, and includes a Mettler Toledo single-use pH and DO sensor, foam sensor, Meissner inlet, two 5-inch exhaust filters, and condenser.
All SUF BPCs are constructed in an ISO 7 (Class 10,000) cleanroom, which complies with cGMP standards.
Features:
• Manufactured with industry-leading CX5-14 film
• C-flex tubing that can be used with tubing welder and various connection options
• Configurable to meet your process needs, including options for single-use sensing (DO, pH, and pressure)
Customize standard BPC systems for optimized single-use technology
Choose from the industry’s largest component library, with over 2,000 unique components that include:
Fittings—straight, reducing, elbow, T-style, X-style, and cross-style
Connectors—quick connectors/disconnectors, aseptic connectors and disconnectors, steam-to and steam-through connectors
Filters—used for bioburden reduction; sterilization grade filters and gas filters
Tubing—thermoplastic elastomers, platinum-cured silicone
Sample ports—septum-style and needle-free ports
Clamps—sliders, pinch, and crimp bands
Especificaciones
Línea de productosHyPerforma™
Cantidad30 L
Condiciones de envíoTemperatura ambiente
Capacidad (métrico)30 L
Unit Size30 L
Contenido y almacenamiento
Almacenar a temperatura ambiente.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documentos y descargas
Certificados
Se muestran 0 resultados, busque arriba un certificado específico
Compartir número de catálogo, nombre o enlace.
1x1 image pixel for data collection