Thermo Scientific™
DynaDrive™ BioProcess Container
Catalog number: SH31193.01
Have Questions?
Thermo Scientific™
DynaDrive™ BioProcess Container
Catalog number: SH31193.01
Have Questions?
Catalog Number
SH31193.01
also known as SH3119301
Unit Size
500 L
Price (USD)
Catalog NumberUnit SizePrice (USD)
SH31193.01
also known as SH3119301
500 LRequest A Quote
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
This bioprocess container (BPC) for the DynaDrive Single-Use Bioreactor (S.U.B.) is made with our proprietary Aegis5-14 film. This film is widely used by many global biopharmaceutical companies for all bioprocessing steps. Aegis5-14 film was introduced in 2013 and has a clean profile, with completed BPOG leachable and extraction (L&E) testing guidelines for all components used in DynaDrive BPCs.
Key benefits
We offer both standard and custom BPC configurations. Standard configurations are for a variety of common applications, while custom configurations are for customer-specific applications. The key benefits for our BPCs include:
  • Enhanced sparging technology to maximize efficiency while delivering improved mass transfer performance
  • The latest sensor technology in single-use or autoclavable probes, including:
    • pH—electrochemical single-use or reusable probes, and optical single-use sensor
    • pO2—optical single-use or reusable probes
    • Foam—single-use foam sensor built into BPC
    • Capacitance—single-use sensor or reusable probe
    • Raman—optical probe port into BPC
  • Optional ports for alternating tangential flow (ATF) or tangential flow filtration (TFF)
  • Built-in condenser system for 3,000 and 5,000 L BPCs, which helps extend the life and reduce the quantity of exhaust filters needed
  • Utilization of crossflow sparging technology to assure proper head-space gas mixing during low-volume seed or terminal cell cultures
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Capacity (Metric)
500 L