| SH31205.11 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID C-Flex, MPU Body and MPU Insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID C-Flex, MPU Body and MPU Insert | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPU Body and MPU Insert | Tubing Material | C-Flex | Unit Size | 1 each |
|
| SH31201.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Unit Size | 1 each |
|
| SH31201.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Unit Size | 1 each |
|
| SH31205.12 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID C-Flex, MPU Body and MPU Insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID C-Flex, MPU Body and MPU Insert | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPU Body and MPU Insert | Tubing Material | C-Flex | Unit Size | 1 each |
|
| SH31201.03 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1/4 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Unit Size | 1 each |
|
| SH31205.13 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 15 ft., 1 in. ID C-Flex, MPU Body and MPU Insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 15 ft., 1 in. ID C-Flex, MPU Body and MPU Insert | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPU Body and MPU Insert | Tubing Material | C-Flex | Unit Size | 1 each |
|
| SH31201.11 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Unit Size | 1 each |
|
| SH31205.01 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, MPU Body and MPU Insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 5 ft., 1 in. ID Silicone, MPU Body and MPU Insert | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPU Body and MPU Insert | Tubing Material | Silicone | Unit Size | 1 each |
|
| SH31201.12 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 3/8 in. ID TPE, 1 1/2 tri-clamp and 3/4 tri-clamp | Unit Size | 1 each |
|
| SH31205.02 | Full specifications | 1 each | Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, MPU Body and MPU Insert | Request A Quote |
Description | Transfer Assembly Jumper Tubing, 10 ft., 1 in. ID Silicone, MPU Body and MPU Insert | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | MPU Body and MPU Insert | Tubing Material | Silicone | Unit Size | 1 each |
|