Transfer Assembly Adapter
Transfer Assembly Adapter
Thermo Scientific™
Transfer Assembly Adapter
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. Simplified connectivity and greater process efficiency for most upstream and downstream process operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SH31322.01Transfer Assembly Adapter, 3/8 to 1/2 in. ID TPE, MPC insert and MPX insert
SH31231.01Transfer Assembly Adapter, 3/8 in. ID TPE, MPC body and and AseptiQuik G connector
SH31375.01Transfer Assembly Adapter, 1 in. ID TPE, AseptiQuik L and G Connectors
SH31232.01Transfer Assembly Adapter, 3/4 to 1/2 in. ID TPE, AseptiQuik L connector and MPX body
SH31428.01Transfer Assembly Adapter, 1 in. ID Silicone, AseptiQuik L and G Connectors
SH31233.01Transfer Assembly Adapter, 1/2 in. ID TPE, 1 1/2 tri-clamp and MPX insert
SH31429.01Transfer Assembly Adapter, 3/4 in. ID Silicone, AseptiQuik L Connector and MPU Body
SH31234.01Transfer Assembly Adapter, 1/2 in. ID TPE, MPX body and 3/4 tri-clamp
SH31430.01Transfer Assembly Adapter, 1/2 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPX Insert
SH31235.01Transfer Assembly Adapter, 3/4 in. ID TPE, 3/4 tri-clamp and AseptiQuik G connector
SH31435.01Transfer Assembly Adapter, 3/8 to 1/2 in. ID Braided Silicone, MPC Insert and MPX Insert
SH31236.01Transfer Assembly Adapter, 3/8 in. ID TPE, MPC insert
SH31436.01Transfer Assembly Adapter, 1/2 in. ID Braided Silicone, MPX Body and 3/4 Tri-Clamp
SH31237.01Transfer Assembly Adapter, 1/2 in. ID TPE, AseptiQuik G connector and MPX body
SH31438.01Transfer Assembly Adapter, 1/2 in. ID Braided Silicone, MPX Insert and 3/4 Tri-Clamp
SH31238.01Transfer Assembly Adapter, 1/2 in. ID TPE, MPX insert and 3/4 tri-clamp
SH31439.01Transfer Assembly Adapter, 3/8 in. ID Braided Silicone, MPC Insert
SH31309.01Transfer Assembly Adapter, 3/4 in. ID TPE, AseptiQuik L connector and MPU body
SH31440.01Transfer Assembly Adapter, 1 in. ID Braided Silicone, 1 1/2 Tri-Clamp and MPU
SH31316.01Transfer Assembly Adapter, 3/4 in. ID TPE, MPU insert and AseptiQuik G connector
SH31228.01Transfer Assembly Adapter, 1 in. ID Silicone, 1 1/2 Tri-Clamp and MPU Insert
SH31241.01Transfer Assembly Adapter, 1/2 in. ID Silicone, AseptiQuik G Connector and MPX Body
SH31226.01Transfer Assembly Adapter, 1 inch ID C-Flex, AseptiQuik L Connector and MPU Body
SH31242.01Transfer Assembly Adapter, 1 in. ID Silicone, MPU Insert and AspetiQuick L Connector
SH31229.01Transfer Assembly Adapter, 1 inch ID TPE, MPU Insert and AseptiQuik L Connector
SH31243.01Transfer Assembly Adapter, 1/2 in. ID Silicone, 1 1/2 Tri-Clamp and MPX Insert
SH31240.01Transfer Assembly Adapter, 1 inch ID TPE, 1 1/2 Tri-Clamp and MPU Insert
SH31425.01Transfer Assembly Adapter, 1/2 in. ID Silicone, MPX Body and 3/4 Tri-Clamp
SH31309.11Transfer Assembly Adapter, 1 inch ID TPE, AseptiQuik L Connector and MPU Body
SH31426.01Transfer Assembly Adapter, 1/2 in. ID Silicone, MPX Insert and 3/4 Tri-Clamp
SH31427.01Transfer Assembly Adapter, 3/8 in. ID Silicone, MPC Insert
SH31420.01Transfer Assembly Adapter, 3/4 in. ID Silicone, 3/4 Tri-Clamp and AseptiQuik G Connector
SH31421.01Transfer Assembly Adapter, 3/8 in. ID Silicone, MPC Body and and AseptiQuik G Connector
SH31422.01Transfer Assembly Adapter, 3/4 in. ID Silicone, MPU Insert and AseptiQuik G Connector
SH31423.01Transfer Assembly Adapter, 3/4 to 1/2 in. ID Silicone, AseptiQuik L Connector and MPX Body
SH31424.01Transfer Assembly Adapter, 3/8 to 1/2 in. ID Silicone, MPC Insert and MPX Insert
SH31431.01Transfer Assembly Adapter, 3/4 in. ID Braided Silicone, 3/4 Tri-Clamp and AseptiQuik G Connector
SH31432.01Transfer Assembly Adapter, 3/8 in. ID Braided Silicone, MPC Body and and AseptiQuik G Connector
SH31433.01Transfer Assembly Adapter, 3/4 in. ID Braided Silicone, MPU Insert and AseptiQuik G Connector
SH31434.01Transfer Assembly Adapter, 3/4 to 1/2 in. ID Braided Silicone, AseptiQuik L Connector and MPX Body
SH31442.01Transfer Assembly Adapter, 1 to 3/4 in. ID Braided Silicone, AseptiQuik L and G Connectors
SH31444.01Transfer Assembly Adapter, 1/2 in. ID Braided Silicone, AseptiQuik G Connector and MPX Body
SH31446.01Transfer Assembly Adapter, 1 in. ID Braided Silicone, AseptiQuik L Connector and MPU Body
SH31447.01Transfer Assembly Adapter, 1 in. ID Braided Silicone, MPU Insert and AseptiQuik L Connector
SH31448.01Transfer Assembly Adapter, 3/4 in. ID Braided Silicone, AseptiQuik L Connector and MPU Body
45 Options
Catalog number SH31322.01
Price (USD)/ 1 each
-
Request A Quote
Description:
Transfer Assembly Adapter, 3/8 to 1/2 in. ID TPE, MPC insert and MPX insert
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Specifications
DescriptionTransfer Assembly Adapter, 3/8 to 1/2 in. ID TPE, MPC insert and MPX insert
Unit Size1 each