| SH31325.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 3/8 in. ID TPE, AseptiQuik G connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 3/8 in. ID TPE, AseptiQuik G connectors | Unit Size | 1 each |
|
| SH31459.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31328.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 1/2 in. ID TPE, AseptiQuik G connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 1/2 in. ID TPE, AseptiQuik G connectors | Unit Size | 1 each |
|
| SH31460.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31331.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 1 in. ID TPE, AseptiQuik L connectors and Y and T connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 1 in. ID TPE, AseptiQuik L connectors and Y and T connectors | Unit Size | 1 each |
|
| SH31463.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y and T Connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y and T Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31337.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 3/4 in. ID TPE, AseptiQuik G connectors and Y and T connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 3/4 in. ID TPE, AseptiQuik G connectors and Y and T connectors | Unit Size | 1 each |
|
| SH31464.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 1 in. ID Silicone, AseptiQuik L Connectors and Y and T Connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 1 in. ID Silicone, AseptiQuik L Connectors and Y and T Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31465.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 3/8 in. ID Braided Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 3/8 in. ID Braided Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31467.01 | Full specifications | 1 each | Transfer Assembly 6-Way Manifold, 1/2 in. ID Braided Silicone, AseptiQuick G Connectors | Request A Quote |
Description | Transfer Assembly 6-Way Manifold, 1/2 in. ID Braided Silicone, AseptiQuick G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|