| SH31327.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 1/2 in. ID TPE, AseptiQuik G connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 1/2 in. ID TPE, AseptiQuik G connectors | Unit Size | 1 each |
|
| SH31324.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 3/8 in. ID TPE, AseptiQuik G connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 3/8 in. ID TPE, AseptiQuik G connectors | Unit Size | 1 each |
|
| SH31449.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31451.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31330.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 1 in. ID TPE, AseptiQuik L connectors and Y and T connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 1 in. ID TPE, AseptiQuik L connectors and Y and T connectors | Unit Size | 1 each |
|
| SH31453.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 1 in. ID Silicone, AseptiQuik L Connectors and Y and T Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 1 in. ID Silicone, AseptiQuik L Connectors and Y and T Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31456.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 3/4 in. ID Silicone, AseptiQuick G Connectors and Y and T Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 3/4 in. ID Silicone, AseptiQuick G Connectors and Y and T Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31457.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 3/8 in. ID Braided Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 3/8 in. ID Braided Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31466.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 1 in. ID Braided Silicone, AseptiQuik L Connectors and Y and T Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 1 in. ID Braided Silicone, AseptiQuik L Connectors and Y and T Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31488.01 | Full specifications | 1 each | Transfer Assembly 5-Way Manifold, 3/4 in. ID Braided Silicone, AseptiQuik G Connectors and Y and T Connectors | Request A Quote |
Description | Transfer Assembly 5-Way Manifold, 3/4 in. ID Braided Silicone, AseptiQuik G Connectors and Y and T Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|