| SH31445.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31323.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 3/8 in. ID TPE, AseptiQuik G connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 3/8 in. ID TPE, AseptiQuik G connectors | Unit Size | 1 each |
|
| SH31441.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31326.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 1/2 in. ID TPE, AseptiQuik G connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 1/2 in. ID TPE, AseptiQuik G connectors | Unit Size | 1 each |
|
| SH31443.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 1 in. ID Silicone, AseptiQuik L Connectors and Y Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 1 in. ID Silicone, AseptiQuik L Connectors and Y Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|
| SH31329.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 1 in. ID TPE, AseptiQuik L connectors and Y connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 1 in. ID TPE, AseptiQuik L connectors and Y connectors | Unit Size | 1 each |
|
| SH31437.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31450.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 3/8 in. ID Braided Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 3/8 in. ID Braided Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31452.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 1/2 in. ID Braided Silicone, AseptiQuik G Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 1/2 in. ID Braided Silicone, AseptiQuik G Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Aseptic | Unit Size | 1 each |
|
| SH31454.01 | Full specifications | 1 each | Transfer Assembly 4-Way Manifold, 1 in. ID Braided Silicone, AseptiQuik L Connectors and Y Connectors | Request A Quote |
Description | Transfer Assembly 4-Way Manifold, 1 in. ID Braided Silicone, AseptiQuik L Connectors and Y Connectors | For Use With (Application) | Monoclonal antibody production, upstream and downstream applications | Quantity | Each | Connector Types | Non-aseptic | Unit Size | 1 each |
|