Thermo Scientific™
Transfer Assembly Y Manifold
Catalog number: SH31458.01
Have Questions?
Thermo Scientific™
Transfer Assembly Y Manifold
Catalog number: SH31458.01
Have Questions?
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
Catalog Number
SH31458.01
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 1/4 in. ID Silicone, AseptiQuik G Connectors and Y Connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 1/4 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 1/8 in. ID TPE, AseptiQuik S connectors and Y connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 1/8 in. ID TPE, AseptiQuik S connectors and Y connector
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 1/8 in. ID TPE, MPC insert, Y connector, and MPC body
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 1/8 in. ID TPE, MPC insert, Y connector, and MPC body
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors and Y Connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik S connectors and Y connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik S connectors and Y connector
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors and Y Connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 1/4 in. ID TPE, AseptiQuik G connectors and Y connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 1/4 in. ID TPE, AseptiQuik G connectors and Y connector
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y Connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik G connectors and Y connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik G connectors and Y connector
Unit Size1 each
Catalog Number
Unit Size
1 each
Description
Transfer Assembly Y Manifold, 1/8 in. ID Silicone, AseptiQuik S Connectors and Y Connector
Price (USD)
Full specifications
DescriptionTransfer Assembly Y Manifold, 1/8 in. ID Silicone, AseptiQuik S Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Showing 10 of 42Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
SH31458.01Full specifications
1 eachTransfer Assembly Y Manifold, 1/4 in. ID Silicone, AseptiQuik G Connectors and Y ConnectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 1/4 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
SH31200.01Full specifications
1 eachTransfer Assembly Y Manifold, 1/8 in. ID TPE, AseptiQuik S connectors and Y connectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 1/8 in. ID TPE, AseptiQuik S connectors and Y connector
Unit Size1 each
SH31200.10Full specifications
1 eachTransfer Assembly Y Manifold, 1/8 in. ID TPE, MPC insert, Y connector, and MPC bodyRequest A Quote
DescriptionTransfer Assembly Y Manifold, 1/8 in. ID TPE, MPC insert, Y connector, and MPC body
Unit Size1 each
SH31458.11Full specifications
1 eachTransfer Assembly Y Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors and Y ConnectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 3/8 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
SH31200.20Full specifications
1 eachTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik S connectors and Y connectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik S connectors and Y connector
Unit Size1 each
SH31458.21Full specifications
1 eachTransfer Assembly Y Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors and Y ConnectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 1/2 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
SH31202.01Full specifications
1 eachTransfer Assembly Y Manifold, 1/4 in. ID TPE, AseptiQuik G connectors and Y connectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 1/4 in. ID TPE, AseptiQuik G connectors and Y connector
Unit Size1 each
SH31458.31Full specifications
1 eachTransfer Assembly Y Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y ConnectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 3/4 in. ID Silicone, AseptiQuik G Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
SH31202.11Full specifications
1 eachTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik G connectors and Y connectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 3/8 in. ID TPE, AseptiQuik G connectors and Y connector
Unit Size1 each
SH31461.01Full specifications
1 eachTransfer Assembly Y Manifold, 1/8 in. ID Silicone, AseptiQuik S Connectors and Y ConnectorRequest A Quote
DescriptionTransfer Assembly Y Manifold, 1/8 in. ID Silicone, AseptiQuik S Connectors and Y Connector
For Use With (Application)Monoclonal antibody production, upstream and downstream applications
QuantityEach
Connector TypesNon-aseptic
Unit Size1 each
Showing 10 of 42Show more
Thermo Scientific Standard Fluid Transfer Assemblies with BioTitan Retention Device increase single-use product flexibility. They connect to our standard Labtainer bioprocess containers (BPCs), Productainer BPCs, single-use bioreactors, mixers, and fermentors right where you need them. They deliver simplified connectivity and enable greater process efficiency for most upstream and downstream process operations.
• One set of stocked products reduces the number of customized stock keeping units (SKUs), simplifies the ordering process, and reduces product storage requirements
• Manufactured with BioTitan Retention Device—designed to provide superior universal tubing retention that minimizes the risk of leakage and failure at connection points
• Multiple manufacturing sites ensure security in product supply
• Produced under cGMP conditions in ISO 7 clean rooms to ensure a high level of quality
• Characterized using ISO, USP, EP, and ASTM test methods to validate performance
Standard Fluid Transfer Assembly products interconnect with Thermo Scientific standard BPC products to deliver a comprehensive and complete set of single-use products to meet the needs of your facility or process. Designed to aid many common production processes such as sampling, transfer, and filling, a fit-for-purpose solution in the required tubing size and length can be easily selected from these designs.
All Standard Fluid Transfer Assembly products are constructed of capped-end connectors and thermoplastic elastomer (TPE) tubing for use with welders. The Extension Set is available in a variety of tubing sizes and lengths to meet your fluid transfer needs.
These Standard Fluid Transfer Assembly products provide a common set of products that addresses an expansive set of needs. They are manufactured at multiple sites globally to minimize concerns of production interruptions and supply shortages. These products are built to our high quality standards to facilitate the success of your process.
Contents & Storage
Room Temperature