Search Thermo Fisher Scientific
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Product Type | Gassing Manifold |
Description | Gassing manifold with 16 outlets |
For Use With (Equipment) | MaxQ 5000 |
Unit Size | Each |
Product Type | Gassing Manifold |
Dimensions (L x W) | 84.5 x 10.2 cm |
Description | Stainless steel shelf for chamber |
For Use With (Equipment) | MaxQ 5000 |
No. of Shelves | 1 |
Unit Size | Each |
Product Type | Gassing Manifold |
Description | Gassing manifold with 8 outlets |
For Use With (Equipment) | MaxQ 5000 |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | For Use With (Equipment) | Price (USD) | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
SHK5000-16GM | Each | Gassing manifold with 16 outlets | MaxQ 5000 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SHK5000-6 | Each | Stainless steel shelf for chamber | MaxQ 5000 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
SHK5000-8GM | Each | Gassing manifold with 8 outlets | MaxQ 5000 | Request A Quote | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Gassing Manifold
Stainless-Steel Shelf