Search Thermo Fisher Scientific
Catalog Number | Description | Electrical Requirements | Speed Range | Temperature Range (Metric) |
---|---|---|---|---|
SHKE4450-1CE | Small Incubated Benchtop Orbital Shaker | 240 V, 50/60 Hz | 15 to 500 rpm, ±1 rpm | Ambient +5° to 80°C |
SHKE4450 | Small Incubated Benchtop Orbital Shaker | 120 V, 50/60 Hz | 15 to 500 rpm, ±1 rpm | Ambient +5° to 80°C |
SHKE4450CC-1CE | Small Incubated Benchtop Orbital Shaker with cooling coil (required chiller) | 240 V, 50/60 Hz | 15 to 500 rpm, ±1 rpm | Ambient -5° to 80°C |
Combine incubated and refrigerated temperatures with a small footprint by using Thermo Scientific™ MaxQ™ 4450 Benchtop Orbital Shakers. These versatile shakers are ideal for incubating a small number of vessels and are used for cell culture, solubility studies, extraction procedures, protein expression and many other applications.
Analog Models:
Digital Models:
Operating Chiller:
Ordering Information:
For a complete system, order platforms and clamps (sold separately).
Requires:
Universal Platform (without clamps) or Dedicated Platform (with pre-installed clamps). All MaxQ 4450 shakers with cooling coil require a chiller sold separately.
Recommended for: