iBind™ Western Device

Catalog number: SLF1000
Have Questions?

iBind™ Western Device

Catalog number: SLF1000
Have Questions?
Catalog Number
SLF1000
Unit Size
1 device
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
4 devices
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SLF10001 device
-
SLF10004PK4 devices
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The iBind Western Device is an automated western-processing device that performs every step from blocking to washes to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source, shakers, or trays.

Learn more about the iBind western systems ›

The iBind Western Device works exclusively with iBind Cards and the iBind Solution Kit or iBind Fluorescent Detection (FD) Solution Kit to process one mini-membrane for immunodetection. The iBind device provides automated convenience and superior reproducibility over traditional manual methods. Each solution is released from the iBind device wells to an iBind Card via SLF in ∼2.5 hours. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions over the membrane, increasing the antigen-antibody interaction.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity
1 Mini Blot
Detection Method
Fluorescence, Chemiluminescent, Chromogenic
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
iBind
Running Time
∼2.5 hr.
Throughput
1 Mini Blot
Type
Western Device
Includes
1 device

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text