Invitrogen™
iBind™ Western Starter Kit
Catalog number: SLF1000S
Have Questions?
Invitrogen™
iBind™ Western Starter Kit
Catalog number: SLF1000S
Have Questions?
Catalog Number
SLF1000S
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SLF1000S1 kit
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The iBind Western Starter Kit includes an iBind Western Device as well as all of the consumables and reagents necessary to get started using this automated western-processing device. The iBind Western Device performs every step in the western protocol from blocking to washes to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source, shakers, or trays.
Learn more about the iBind western systems ›
The iBind Western Device works exclusively with iBind Cards and the iBind Solution Kit to process one mini-membrane for immunodetection. The iBind device provides automated convenience and superior reproducibility over traditional manual methods. Each solution is released from the iBind device wells to an iBind Card via SLF in ∼2.5 hours. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions over the membrane, increasing the antigen-antibody interaction.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Capacity
1 mini blot (8 x 8 cm)
Run Time
∼2.5 hours
Detection Method
Chromogenic, Chemiluminescent, Fluorescent
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
iBind™
Product Type
Starter Kit
Contents & Storage
• 1 iBind™ Western Device
• 10 iBind™ Cards (store at room temperature)
• 1 iBind™ Solution Kit (store at 2–8°C)