Invitrogen™

iBind™ Flex Western Device

Catalog number: SLF2000
Have Questions?
Invitrogen™

iBind™ Flex Western Device

Catalog number: SLF2000
Have Questions?
Catalog Number
SLF2000
Unit Size
1 device
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
2 devices
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SLF20001 device
-
SLF20002PK2 devices
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The iBind Flex Western Device is an automated western-processing device that performs every step from blocking to washes to antibody incubations via sequential lateral flow (SLF). SLF allows the timely release and flow of solutions and antibodies to the membrane without need of an external power source, shakers, or trays. Each solution is released from the iBind Flex device wells to an iBind Flex Card via SLF; the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

Learn more about the iBind western systems ›

The iBind Flex western detection procedure minimizes hands-on time and is highly reproducible. Setup time is fast: within 2.5 h you are ready for the detection step. Compared to manual methods, the iBind Flex western system requires less primary antibody, enables reproducible results, and is compatible with downstream detection protocols.

The iBind Flex Western Device includes three user-interchangeable well inserts to process up to one midi blot, two mini blots, or six vertically-cut strip blots when used with the iBind Flex Cards and iBind Flex Solution Kit or iBind Flex Fluorescent Detection (FD) Solution Kit.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

Capacity
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Detection Method
Fluorescence, Chemiluminescent, Chromogenic
For Use With (Application)
Western Blot
Membrane Compatibility
Nitrocellulose, PVDF
Product Line
iBind Flex
Running Time
∼2.5 hr.
Throughput
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots
Type
Western Device
Includes
Roller
Shipping Condition
Room Temperature

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text