Invitrogen™

iBind™ Flex Solution Kit

Catalog number: SLF2020
Have Questions?
Invitrogen™

iBind™ Flex Solution Kit

Catalog number: SLF2020
Have Questions?
Catalog Number
SLF2020
Unit Size
1 kit
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
1 kit,4 kits
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SLF20201 kit
-
SLF2020X41 kit,4 kits
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The iBind Flex Solution Kit is intended for use with the iBind Flex Western Device, which is an automated western-processing device that performs every step in the western workflow from blocking to washes to antibody incubations. The iBind solutions act as the blocking, washing, and dilution solutions in the iBind immunodetection process and have been optimized for ideal viscosity. Each solution is released from the iBind Flex device wells to the iBind Flex Card through sequential lateral flow (SLF); the solutions are then wicked towards the stack region of the card. The glass fiber matrix of the card allows for homogenous and consistent flow of the solutions to the membrane, increasing the antigen-antibody interaction.

Learn more about the iBind western systems ›

The iBind Flex Solutions Kit, together with iBind Flex Western Device and iBind Flex Cards, bring automation and simplicity to the western protocol. Compared to manual methods, the iBind Flex western system requires less primary antibody, enables reproducible results, and is compatible with downstream detection protocols. Each iBind Flex Solution Kit contains sufficient solution for 10 midi or 20 mini blots. For fluorescent applications use the iBind Flex Fluorescent Detection (FD) Solution Kit.

For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
iBind Flex Western Device
Kit Contents
iBind™ Flex 5X buffer, iBind™ Flex 100X additive
No. of Reactions
10 midi blots or 20 mini blots
Shipping Condition
Approved for shipment at Room Temperature or on Wet Ice
Detection Method
Colorimetric, Chromogenic, Chemiluminescent
Membrane Compatibility
PVDF, Nitrocellulose
Product Line
iBind™
Product Type
Flex Solution Kit

Contents & Storage

• iBind Flex 5X Buffer
• iBind Flex 100X Additive

Store at 2–8°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text