Lambda DNA/HindIII Marker
Lambda DNA/HindIII Marker
Thermo Scientific™
Lambda DNA/HindIII Marker
Thermo Scientific Lambda DNA/HindIII Marker is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNARead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SM01015 x 50 μg
SM010225 x 50 μg
Catalog number SM0101
Price (USD)
107.65
Online Exclusive
117.00
Save 9.35 (8%)
5 x 50 µg
Add to cart
Quantity:
5 x 50 μg
Request bulk or custom format
Price (USD)
107.65
Online Exclusive
117.00
Save 9.35 (8%)
5 x 50 µg
Add to cart
Ask our AI about this Product
Thermo Scientific Lambda DNA/HindIII Marker is recommended for sizing of linear double-stranded large DNA fragments in agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
The marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively, and it can be labeled radioactively or non-radioactively with Klenow Fragment or Klenow Fragment, exo- with the fill-in reaction (see Resources for Protocols).
Highlights
• Sizing and approximate quantification of large DNA fragments
• Sharp bands
• Supplied with loading dye for sample DNA
Note
The cohesive ends (the 12 nt cos site of lambda DNA) of fragments indicated may anneal and form additional bands. These fragments can be separated by heating at 65°C for 5 minutes and then cooling on ice for 3 minutes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityAgarose Gels
Product TypeDNA/Hind III Marker
Quantity5 x 50 μg
Ready to LoadNo
Gel TypeAgarose
Unit Size5 x 50 µg
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
What is the recommended voltage for electrophoresis with Thermo Scientific DNA and RNA ladders? What does V/cm mean?
Each of Thermo Scientific DNA and RNA ladders has a recommended voltage given in V/cm. Please refer to the gel picture in the product information sheet. V/cm stands for the amount of voltage recommended per cm of the distance between the two electrodes of the gel box (not gel length).
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link