Lambda DNA/HindIII Marker, ready-to-use
Lambda DNA/HindIII Marker, ready-to-use
Thermo Scientific™
Lambda DNA/HindIII Marker, ready-to-use
Thermo Scientific Lambda DNA/HindIII Marker, ready-to-use is recommended for sizing of linear double-stranded large DNA on agarose gels. Lambda DNARead more
Have Questions?
Catalog NumberQuantity
SM01035 x 50 μg
Catalog number SM0103
Price (EUR)
99,00
5 x 50 µg
-
Add to cart
Quantity:
5 x 50 μg
Request bulk or custom format
Price (EUR)
99,00
5 x 50 µg
Add to cart
Ask our AI about this Product
Thermo Scientific Lambda DNA/HindIII Marker, ready-to-use is recommended for sizing of linear double-stranded large DNA on agarose gels. Lambda DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme(s) and purified and dissolved in storage buffer. DNA fragments contain various sticky ends depending on the restriction enzyme used for preparation of the marker.
Lambda DNA/HindIII Marker, ready-to-use is premixed with 6X DNA Loading Dye for direct loading (after heating) onto agarose gels.
Highlights
• Sizing and approximate quantification of large DNA fragments
• Sharp bands
• Premixed with 6X DNA Loading Dye for a direct loading (after heating) and for room temperature storage
• Supplied with loading dye for sample DNA
Note
The cohesive ends (the 12 nt cos site of lambda DNA) of fragments indicated may anneal and form additional bands. These fragments can be separated by heating at 65°C for 5 minutes and then cooling on ice for 3 minutes.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel CompatibilityAgarose Gels
Product TypeDNA/Hind III Marker
Quantity5 x 50 μg
Ready to LoadYes
Gel TypeAgarose
Unit Size5 x 50 µg
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link