Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:These products are now shipping under ambient conditions. Learn more.
Thermo Scientific™
pUC19 DNA/MspI (HpaII) Marker, ready-to-use
Catalog number: SM0223
Have Questions?
Thermo Scientific™
pUC19 DNA/MspI (HpaII) Marker, ready-to-use
Catalog number: SM0223
Have Questions?
Catalog Number
SM0223
Unit Size
50 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SM022350 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific pUC19 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation. pUC19 DNA/MspI (HpaII) Marker, ready-to-use, is premixed with 6X DNA Loading Dye for direct loading onto agarose and polyacrylamide gels.
Highlights
• Sizing and approximate quantification of small DNA fragments
• Sharp bands
• Premixed with 6X DNA Loading Dye for direct loading and room temperature storage
• Supplied with loading dye for sample DNA
Note
• Bands at 501 bp and 498 bp form an anomalous pattern on polyacrylamide gels.
• The shortest fragments (oblique) are not visible in standard electrophoresis.
• Fragment lengths are predicted by computer analysis of the respective DNA sequences.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration
6X
Gel Type
Agarose
Product Type
DNA/MSPI (HPAII) Marker
Ready to Load
Yes
Gel Compatibility
Agarose Gels
Green Features
Sustainable packaging