Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Product Type | Quantity | Size Range |
---|---|---|---|
SM0243 | DNA Ladder | 50 μg, 100 applications | 100 to 1000 bp |
SM1213 | Ultra Low Range DNA Ladder | 50 μg | 10 to 300 bp |
SM1193 | Low Range DNA Ladder | 50 μg | 25 to 700 bp |
SM0373 | DNA Ladder | 50 μg | 50 to 1000 bp |
SM1334 | DNA Ladder | 50 μg | 75 to 20000 bp |
SM0323 | DNA Ladder | 50 μg | 100 to 3000 bp |
SM0334 | DNA Ladder Mix | 50 μg, 100 applications | 100 to 1000 bp |
SM0314 | DNA Ladder | 50 μg, 100 applications | 250 to 10000 bp |
SM1353 | High Range DNA Ladder | 50 μg | 10171 to 48502 bp |
SM1333 | DNA Ladder | 500 Applications | 75 to 20000 bp |
SM0244 | DNA Ladder | 5 x 50 μg, 500 applications | 100 to 1000 bp |
SM0324 | DNA Ladder | 5 x 50 μg, 500 applications | 100 to 3000 bp |
SM0333 | DNA Ladder Mix | 5 x 50 μg, 500 applications | 100 to 1000 bp |
SM0313 | DNA Ladder | 5 x 50 μg, 500 applications | 250 to 10000 bp |
Thermo Scientific GeneRuler DNA Ladders serve as precise molecular weight markers for the sizing and approximate quantification of double-stranded DNA fragments, accommodating a range from 10 bp up to 48.5 kb. The uniform intensity of the bands, coupled with distinct bright reference bands, facilitates accurate sizing and orientation during gel electrophoresis. Furthermore, the defined quantity of DNA within each band enables the approximate quantification of experimental DNA samples.
GeneRuler DNA ladders are provided in a convenient ready-to-use format incorporating 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G). The three dye present in the buffer allow for easy tracking of DNA migration during electrophoresis.
Highlights
Sustainability features
Ready-to-use GeneRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Share catalog number, name or link