phiX174 DNA/BsuRI (HaeIII) Marker
These products are now shipping under ambient conditions. Learn more.
phiX174 DNA/BsuRI (HaeIII) Marker
Thermo Scientific™
phiX174 DNA/BsuRI (HaeIII) Marker
Green features
Thermo Scientific phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved inRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SM02525 x 50 μg
SM025150 μg
Catalog number SM0252
Price (USD)
375.65
Online Exclusive
477.00
Save 101.35 (21%)
5 x 50 µg
-
Add to cart
Quantity:
5 x 50 μg
Request bulk or custom format
Price (USD)
375.65
Online Exclusive
477.00
Save 101.35 (21%)
5 x 50 µg
Add to cart
Ask our AI about this Product
Thermo Scientific phiX174 DNA is digested to completion with the appropriate Thermo Scientific restriction enzyme, then purified and dissolved in storage buffer. The DNA fragments contain blunt or sticky ends, depending on the restriction enzyme used for the markers preparation.
Thermo Scientific phiX174 DNA/BsuRI (HaeIII) Marker can be labeled radioactively with T4 Polynucleotide Kinase. Alternatively it can be labeled radioactively or non-radioactively with Klenow fragment or Klenow Fragment, exo- with the fill-in reaction, see protocols in Resources.
Highlights
• Sizing and approximate quantification of small DNA fragments
• Sharp bands
• Supplied with loading dye for sample DNA
Note
• Bands at 310, 281, and 271 bp form an anomalous pattern on polyacrylamide gels.
• The shortest fragments (oblique) are not visible in standard electrophoresis.
• Fragment lengths are predicted by computer analysis of the respective DNA sequences.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LinephiX174 DNA/BsuRI (HaeIII) Marker
Product TypeDNA/BSURI (HAEIII) Marker
Quantity5 x 50 μg
Ready to LoadNo
Gel TypeAgarose
Unit Size5 x 50 µg
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link