GeneRuler Low Range DNA Ladder
Inquire about OEM or Commercial Supply version of this product here.
In April 2018 this product was upgraded to include TriTrack DNA Loading Dye for better visualization.
GeneRuler Low Range DNA Ladder
Thermo Scientific™
GeneRuler Low Range DNA Ladder
Green features
Thermo Scientific GeneRuler Low Range DNA Ladder is recommended for sizing and approximate quantification of a double-stranded DNA in theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
SM119150 μg
SM11925 x 50 μg
Catalog number SM1191
Price (USD)/ 50 µg
82.65
Online exclusive
90.50 
Save 7.85 (9%)
-
Add to cart
Quantity:
50 μg
Request bulk or custom format
Price (USD)/ 50 µg
82.65
Online exclusive
90.50 
Save 7.85 (9%)
Add to cart
GeneRuler Low Range DNA Ladder
Catalog numberSM1191
Price (USD)/ 50 µg
82.65
Online exclusive
90.50 
Save 7.85 (9%)
-
Add to cart
Thermo Scientific GeneRuler Low Range DNA Ladder is recommended for sizing and approximate quantification of a double-stranded DNA in the range of 25 bp to 700 bp on agarose or polyacrylamide gels. The DNA ladder consists of 10 DNA fragments and is provided with 6X TriTrack DNA Loading Dye.
Highlights of Low Range DNA Ladder
Easy visualization—three-color tracking of DNA migration during electrophoresis with 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G)
Simplicity—one loading dye for a variety of samples
Sharp bands—clear results with mixtures of individual chromatography-purified DNA fragments
Product use
This double-stranded DNA ladder is compatible with 3% standard and precast agarose slab gels and can be visualized after ethidium bromide or SYBR Safe staining. A uniform intensity of bands and bright reference bands allow easy sizing and orientation. A defined amount of DNA in each band enables approximate quantification of sample DNA.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration6X
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Bromophenol Blue, Orange G
Product TypeDNA Ladder
Ready to LoadNo
Size Range25 to 700 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineGeneRuler
Quantity50 μg
Unit Size50 µg