FastRuler Ultra Low Range DNA Ladder, ready-to-use
These products are now shipping under ambient conditions. Learn more.
Inquire about OEM or Commercial Supply version of this product here.
FastRuler Ultra Low Range DNA Ladder, ready-to-use
Thermo Scientific™
FastRuler Ultra Low Range DNA Ladder, ready-to-use
Green features
Thermo Scientific FastRuler Ultra Low Range DNA Ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNARead more
Have Questions?
Catalog number SM1233
Price (USD)/ 2 x 500 µL
90.76
Special Offer
Online exclusive
Ends: 16-Dec-2024
105.00 
Save 14.24 (14%)
-
Add to cart
Request bulk or custom format
Price (USD)/ 2 x 500 µL
90.76
Special Offer
Online exclusive
Ends: 16-Dec-2024
105.00 
Save 14.24 (14%)
Add to cart
FastRuler Ultra Low Range DNA Ladder, ready-to-use
Catalog numberSM1233
Price (USD)/ 2 x 500 µL
90.76
Special Offer
Online exclusive
Ends: 16-Dec-2024
105.00 
Save 14.24 (14%)
-
Add to cart
Thermo Scientific FastRuler Ultra Low Range DNA Ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.
The ladder is especially useful for electrophoresis of PCR products.
FastRuler Ultra Low Range DNA Ladder, ready-to-use contains dephosphorylated DNA fragments and is ideal for forward 5-end labeling with T4 Polynucleotide Kinase (see the Certificate of Analysis for the detailed labeling protocol).
Highlights
• Fast separation (8 to 14 min)
• Short separation distance (10 to 20 mm)
• Sharp bands, easy-to-remember fragment sizes and quantities
• Ready-to-use—premixed with loading dye
• Supplied with loading dye solution for sample DNA
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel TypeAgarose
Label or DyeXylene Cyanol FF, Orange G
Product TypeDNA Ladder
Ready to LoadYes
Shelf Life6 Months
Size Range10 to 200 bp
Gel CompatibilityAgarose Gels
Green FeaturesSustainable packaging
Product LineFastRuler
Quantity2 x 500 μL
Unit Size2 x 500 µL