Informational:Inquire about OEM or Commercial Supply version of this product here.
Informational:These products are now shipping under ambient conditions. Learn more.
Thermo Scientific™

ZipRuler Express DNA Ladder Set, ready-to-use

Catalog number: SM1373
Have Questions?
Thermo Scientific™

ZipRuler Express DNA Ladder Set, ready-to-use

Catalog number: SM1373
Have Questions?
Catalog Number
SM1373
Unit Size
2 x 50 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SM13732 x 50 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific ZipRuler Express DNA Ladder Set contains two ladders: ZipRuler Express DNA Ladder 1 and ZipRuler Express DNA Ladder 2. Both are mixtures of chromatography-purified individual DNA fragments.The set is designed for fast and highly accurate sizing of a broad range (100-20,000 bp) of DNA fragments and their approximate quantification under a variety of electrophoresis conditions. The band pattern is not affected by the composition of electrophoresis buffer, voltage or gel percentage (see Table 1).

Highlights

• Flexible—can be run in two lanes or combined in one lane
• Composed of chromatography purified individual DNA fragments
• Ready-to-use—premixed with a loading dye for direct loading and room temperature storage
• Supplied with loading dye for sample DNA

Features
• For fast separation, the ZipRuler Express DNA Ladder 1 and 2 are loaded into two different wells in the same gel to evaluate a wide range of DNA bands during short electrophoresis runs. For longer runs, equal amounts of both ladders are loaded into the same gel well. Three reference bands will then be formed during electrophoresis of the mixed ZipRuler ladder. Longer electrophoresis run times are needed to achieve full separation of complex ladder.

ZipRuler Express DNA Ladder 1 is premixed with the 6X Orange DNA Loading Dye (orange G and xylene cyanol), whereas ZipRuler Express DNA Ladder 2 is premixed with the 6X MassRuler DNA Loading Dye (bromophenol blue).

When both ladders are loaded in the same gel lane, the migration of DNA is monitored by three electrophoresis tracking dyes (bromophenol blue, xylene cyanol FF and orange G).

The set is supplied with 6X Orange DNA Loading Dye for sample DNA.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
6X
Gel Type
Agarose
Label or Dye
Xylene Cyanol FF, Orange G, Bromophenol Blue
Product Type
DNA Ladder Set
Ready to Load
Yes
Shelf Life
6 Months
Size Range
100 to 20000 bp
Gel Compatibility
Agarose Gels
Green Features
Sustainable packaging
Product Line
ZipRuler

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text