Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™
NoLimits 8000 bp DNA Fragment
Catalog number: SM1521
Have Questions?
Thermo Scientific™
NoLimits 8000 bp DNA Fragment
Catalog number: SM1521
Have Questions?
Catalog Number
SM1521
Unit Size
10 µg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SM152110 µg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.Plasmid DNA, digested with Thermo Scientific Restriction Enzymes, is chromatography-purified to prepare the individual DNA fragments from the digestion mixture. The results are extremely pure DNA fragments, free of any truncated or degraded molecules. These custom DNA fragments are the best custom standards for applications ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Features
• Broad range: 10-20,000 bp
• Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
• Precise DNA concentration
• Chromatography-purified individual DNA fragments
Applications
• Gel electrophoresis
• Capillary electrophoresis
• DNA quantification
• HPLC analysis
• Applications where a custom DNA size standard is required.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel Type
Agarose
Product Type
DNA Fragment
Ready to Load
No
Size Range
10 to 20000 bp
Gel Compatibility
Agarose Gels
Green Features
Sustainable packaging
Product Line
NoLimits