Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

RiboRuler High Range RNA Ladder, ready-to-use

Catalog number: SM1823
Have Questions?
Thermo Scientific™

RiboRuler High Range RNA Ladder, ready-to-use

Catalog number: SM1823
Have Questions?
Catalog Number
SM1823
Unit Size
5 x 40 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SM18235 x 40 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific RiboRuler High Range RNA Ladder 200 to 6000 bases, ready-to-use is a mixture of 8 chromatography purified single-stranded RNA transcripts (200 to 6000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments. Ladder is premixed with loading dye.

RiboRuler High Range RNA Ladder, ready-to-use, is composed of chromatography-purified transcripts that makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically. To minimize the possibility of contamination during the use, the ladder is aliquotted in smaller volumes.

Highlights

  • Sharp bands of uniform intensity
  • Easy-to-remember band sizes and quantities
  • Predetermined concentration of ladder bands allows for approximate RNA quantification
  • Supplied with 2X RNA Loading Dye

Applications

  • RNA sizing and quantification on native or denaturing gels
  • Northern blotting
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
2X
Gel Type
Agarose
Label or Dye
Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide
Product Type
High Range DNA Ladder
Size Range
200 to 6000 bp
Product Line
RiboRuler

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text