Informational:Inquire about OEM or Commercial Supply version of this product here.
Thermo Scientific™

RiboRuler Low Range RNA Ladder, ready-to-use

Catalog number: SM1833
Have Questions?
Thermo Scientific™

RiboRuler Low Range RNA Ladder, ready-to-use

Catalog number: SM1833
Have Questions?
Catalog Number
SM1833
Unit Size
5 x 40 µL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SM18335 x 40 µL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Thermo Scientific RiboRuler Low Range RNA Ladder 100-1000 bp, ready-to-use, is a mixture of 7 chromatography purified single-stranded RNA transcripts (100–1000 bp long), produced from specific templates that contain a fragment of the pTZ19R polylinker and Lambda phage fragments.

This ladder is premixed with loading dye.

RiboRuler Low Range RNA Ladder, ready-to-use, is composed of chromatography-purified transcripts what makes them free from NTPs and RNA degradation products. The concentration of each RNA transcript is determined spectrophotometrically. To minimize the possibility of contamination during the use the ladder is aliquotted in smaller volumes.

RiboRuler Low Range RNA Ladder is optimal for end-labeling using T4 Polynucleotide Kinase (see protocol in the Resources) and is ideal for Northern blots.

Highlights

  • Sharp bands of uniform intensity
  • Easy-to-remember band sizes and quantities
  • Approximate RNA quantification by predetermined concentration of ladder bands
  • Supplied with 2X RNA Loading Dye

Applications

  • RNA sizing and quantification on native or denaturing gels
  • Northern blotting.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Concentration
2X
Gel Type
Agarose
Label or Dye
Xylene Cyanol FF, Bromophenol Blue, Ethidium Bromide
Product Type
DNA Ladder
Size Range
100 to 1000 bp
Product Line
RiboRuler

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text