Search Thermo Fisher Scientific
FIGURE: 1 / 1
This Mouse TNFalpha Standard is calibrated against NIBSC Mouse TNFalpha Reference Standard lot (8)8/532. 1 Thermo pg = 0.71 NIBSC pg = 0.14 NIBSC unit.
Lyophilized product is shipped on cold packs. Stable until expiration if stored desiccated at -20°C. Reconstituted cytokine standards should be stored in aliquots at -70°C. Each aliquot should be used only once and discarded afterwards. Stability of reconstituted and frozen cytokine standards may vary. Controls should be run with each standard curve.
Sufficient For: ELISA standard curve with replicates or as a control in Western blots.
2450 pg/ml when reconstituted at the recommend volume.
Reconstitute with distilled water at volume indicated on label. Use or freeze product within 1 hour of reconstitution.