Purified from Streptomyces avidinii and provided in a phosphate buffered saline solution. The streptavidin/avidin-biotin staining and detection strategy is usedRead more
Have Questions?
Catalog number SNN1004
Price (USD)/ 1 mg
245.00
-
Add to cart
Price (USD)/ 1 mg
245.00
Add to cart
Streptavidin HRP
Catalog numberSNN1004
Price (USD)/ 1 mg
245.00
-
Add to cart
Purified from Streptomyces avidinii and provided in a phosphate buffered saline solution. The streptavidin/avidin-biotin staining and detection strategy is used when increased assay sensitivity is desired for a wide variety of systems.
Format/Formulation: HRP Conjugate
For Laboratory Use
Specifications
Concentration2 mg/mL
ConjugateHRP
Quantity1 mg
Shipping ConditionApproved for shipment on Wet or Dry Ice