Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
SO100 | 10 µM | 82.50 | - |
Thermo Scientific M13/pUC sequencing primers anneal to the region in the 5'-terminus of the lacZ gene. All primers are supplied as 10 µM aqueous solutions.
Applications
• Sequencing of DNA fragments inserted into the MCS within the lacZ gene of various cloning vectors, such as pUC19, pTZ19R, pTZ57R, M13mp18, and pBluescript II
• Colony screening by PCR