M13/pUC Sequencing Primer (-40), 17-mer
This product is being discontinued effective 30 September 2025. Please contact your local sales reptesentative or our customer care teams for additional assistance.
M13/pUC Sequencing Primer (-40), 17-mer
Thermo Scientific™

M13/pUC Sequencing Primer (-40), 17-mer

Thermo Scientific M13/pUC Sequencing Primer (-40), 17-mer is a synthetic single-stranded 17-mer oligonucleotide with free 5'- and 3'-hydroxyl ends. ThisRead more
Have Questions?
Catalog NumberQuantity
SO11310 μM, 60 μL
Catalog number SO113
Price (EUR)
58,50
10 µM
-
Add to cart
Quantity:
10 μM, 60 μL
Price (EUR)
58,50
10 µM
Add to cart
Ask our AI about this Product

Thermo Scientific M13/pUC Sequencing Primer (-40), 17-mer is a synthetic single-stranded 17-mer oligonucleotide with free 5'- and 3'-hydroxyl ends. This M13/pUC primer anneals to the region in the 5'-terminus of the lacZ gene. All primers are supplied as 10 μM aqueous solutions.

Applications
• Sequencing of DNA fragments inserted into the MCS within the lacZ gene of various cloning vectors, such as pUC19, pTZ19R, pTZ57R, M13mp18, and pBluescript II
• Colony screening by PCR

Primer Sequence: 5'-d(GTTTTCCCAGTCACGAC)-3'

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
FormLiquid
Product TypeSequencing Primer
Quantity10 μM, 60 μL
Shipping ConditionDry Ice
VectorpUC19, pTZ19R, pTZ57R, M13mp18, pBluescript II
Concentration10 μM
PrimerM13
Unit Size10 µM
Contents & Storage
M13/pUC Sequencing Primer (-40), 17-mer, 10 μM (60 μL)

Store at –20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link