Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Quantity |
---|---|
SO116 | 10 μM, 56 μL |
Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends. The SP6 Promoter Sequencing Primer, 18-mer is complementary to the SP6 RNA Polymerase promoter region and is supplied as 10 μM aqueous solutions.
Applications
• Sequencing of DNA fragments located downstream from the SP6 RNA polymerase promoter sequence in common cloning vectors, such as pTZ19R, pTZ57R, and pBluescript II
Promoter Sequence 5'-d (ATTTAGGTGACACTATAG)-3'
Related products
SP6 promoter Sequencing Primer, 24-mer
T7 promoter Sequencing Primer, 20-mer
T3 promoter Sequencing Primer, 17-mer
T3 promoter Sequencing Primer, 24-mer
Notes
• Primers cannot be used for certain plasmids that contain truncated, but still fully functional promoters
• Primers are not phosphorylated
Share catalog number, name or link