Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Unit Size | Price (USD) | Availability | Quantity | |
---|---|---|---|---|---|
SO117 | 10 µM | 84.25 | - |
Thermo Scientific Transcription Promoter Sequencing Primers are single-stranded oligonucleotides with 5'- and 3'-hydroxyl ends. This SP6 Promoter Sequencing Primer, 24-mer is complementary to the SP6 RNA Polymerase promoter region and is supplied as 10 μM aqueous solutions.
Applications
• Sequencing of DNA fragments located downstream from the SP6 RNA polymerase promoter sequence in common cloning vectors, such as pTZ19R, pTZ57R, and pBluescript II
Promoter Sequence: 5'-d (CATACGATTTAGGTGACACTATAG)-3'
Related products
SP6 promoter Sequencing Primer, 18-mer
T7 promoter Sequencing Primer, 20-mer
T3 promoter Sequencing Primer, 17-mer
T3 promoter Sequencing Primer, 24-mer
Notes
• Primers cannot be used for certain plasmids that contain truncated, but still fully functional promoters
• Primers are not phosphorylated