Oligo(dT)18 Primer
Oligo(dT)<sub>18</sub> Primer
Thermo Scientific™

Oligo(dT)18 Primer

Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as深入閱讀
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
SO13160 μL
SO132120 μL
產品號碼 SO131
價格 (TWD)
2,800.00
線上優惠
Ends: 30-Sep-2025
3,480.00
您節省 680.00 (20%)
60 µL
新增至購物車
Quantity:
60 μL
價格 (TWD)
2,800.00
線上優惠
Ends: 30-Sep-2025
3,480.00
您節省 680.00 (20%)
60 µL
新增至購物車
Ask our AI about this Product
Thermo Scientific Oligo(dT)18 Primer is a synthetic single-stranded 18-mer oligonucleotide with 5'- and 3'-hydroxyl ends. The primer is supplied as a ready-to-use, 20X concentrated aqueous solution.

Applications
• cDNA Synthesis
• RT-qPCR
• RT-PCR

Related products
Anchored Oligo dT
Random Hexamer Primer
For Research Use Only. Not for use in diagnostic procedures.
規格
For Use With (Application)cDNA Synthesis
FormLiquid
Primer Length18-mer
Product TypePrimer
Quantity60 μL
Shipping ConditionDry Ice
Concentration0.5 μg/μL
PrimerOligo dT
Unit Size60 µL
內容物與存放
• Oligo(dT)18 Primer (60 μL at 0.5 μg/μL)

Store at –20°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

文件與下載

證書

安全資料表

分享目錄編號、產品名稱或連結。