Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 Integrated SpeedVac™ Systems and kits. Fully integrated for fast installation, programming. Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 System is designed for medium capacity (i.e.,120 x 1.5 mL tubes, other formats). SpeedVac SPD2030 System allows higher capacity (i.e.,200 x 1.5 mL tubes, other formats).
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Vacuum concentrator only
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
146 lb.
Shipping Weight (Metric)
86 kg
Model
SPD1030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor and accessories- order separately)
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor and accessories- order separately)
Electrical Requirements
230 V 50 Hz
Includes
Vacuum concentrator only
Price (USD)
17,400.00
Availability
-
Full specifications
Quantity
Certifications
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Vacuum concentrator only
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
198 lb.
Shipping Weight (Metric)
90 kg
Model
SPD1030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor and accessories- order separately)
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Diaphragm pump, cold trap and vacuum gauge
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
198 lb.
Shipping Weight (Metric)
90 kg
Model
SPD1030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor- order separately)
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Diaphragm pump, cold trap and vacuum gauge
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
250 lb.
Shipping Weight (Metric)
113 kg
Model
SPD2030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD2030 Integrated Vacuum Concentrator (rotor- order separately)
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Diaphragm pump, cold trap and vacuum gauge
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
250 lb.
Shipping Weight (Metric)
113 kg
Model
SPD2030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD2030 Integrated Vacuum Concentrator (rotor- order separately)
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Vacuum concentrator only
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
146 lb.
Shipping Weight (Metric)
86 kg
Model
SPD1030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor and accessories- order separately)
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor and accessories- order separately)
230 V 50 Hz
Vacuum concentrator only
17,400.00
-
Certifications
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Vacuum concentrator only
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
198 lb.
Shipping Weight (Metric)
90 kg
Model
SPD1030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor and accessories- order separately)
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Diaphragm pump, cold trap and vacuum gauge
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
198 lb.
Shipping Weight (Metric)
90 kg
Model
SPD1030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD1030 Integrated Vacuum Concentrator (rotor- order separately)
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 120 x 1.5 mL tubes, 10 x 15 mL tubes, 6 x 50 mL tubes and 2 microwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
UL Listed to US and Canadian requirements. CE marked.This product is not available for sale in California. Please contact Customer Service for possible alternatives.
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Diaphragm pump, cold trap and vacuum gauge
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
250 lb.
Shipping Weight (Metric)
113 kg
Model
SPD2030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD2030 Integrated Vacuum Concentrator (rotor- order separately)
UL Listed to US and Canadian requirements. CE marked
Holds
Accommodates up to 200 x 1.5 mL tubes, 30 x 15 mL tubes, 48 x 50 mL tubes, 20 micro well plates and 8 Deepwell plates. Other rotor options available to accommodate a variety of tubes, vials and plates.
Includes
Diaphragm pump, cold trap and vacuum gauge
Max. Vacuum
<10 Torr (13 mbar, 1.3k Pa)
Shipping Weight (English)
250 lb.
Shipping Weight (Metric)
113 kg
Model
SPD2030
Temperature Range (Metric)
Ambient to 80°C
Chamber Material
PTFE-coated Aluminum
Description
SpeedVac SPD2030 Integrated Vacuum Concentrator (rotor- order separately)
Electrical Requirements
230 V 50 Hz
Amperage
8 A
Weight (Metric)
94 kg
Dimensions (L x W x H)
31 x 28 x 19 in. (79 x 71 x42 cm)
Unit Size
Each
Showing 10 of 12Show more
Easy to Use:
Plug and play installation: Integrated solution includes a concentrator, oil-free vacuum pump and cold trap all integrated in one unit. No tubing connections needed.
Intuitive and user friendly program digital display which allows use of three preset programs and ability to create three customizable programs to set heat, run time, ramp rate and vacuum level for efficient operation.
Wide variety of rotors to support sample format from micro tubes, vials and microplates to 250-ml flasks
Audible alarm to alert user when operation is finished
Kits are available, which includes the integrated system and rotor
Robust, Low-Maintenance Design:
Oil-free vacuum pump is virtually maintenance free
PTFE coated chamber protects the chamber from corrosion, while making cleaning easy
Enhanced Evaporation:
Faster sample drying provided with radiant cover
Vacuum ramping and pressure controls eliminate sample bumping to maintain sample integrity
Reduced vapor path from concentrator to refrigerated vapor trap accelerates evaporation
Rotors fit a wide variety of sample formats such as tubes, flasks, and shallow-well and deep-well microplates