Piko PCR Plate, 24-well, clear
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Piko PCR Plate, 24-well, clear
Thermo Scientific™

Piko PCR Plate, 24-well, clear

The Thermo Scientific 24-well Piko PCR Plates, clear,belong to a family of PCR plates which have the footprint of aRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorNo. of Wells
SPL024050Clear24
SPL0240Clear24
2 Options
Catalog number SPL024050
Price (USD)/ 50 plates
135.65
Online exclusive
152.00 
Save 16.35 (11%)
-
Add to cart
Color:
Clear
No. of Wells:
24
Price (USD)/ 50 plates
135.65
Online exclusive
152.00 
Save 16.35 (11%)
Add to cart
Piko PCR Plate, 24-well, clear
Catalog numberSPL024050
Price (USD)/ 50 plates
135.65
Online exclusive
152.00 
Save 16.35 (11%)
-
Add to cart
The Thermo Scientific 24-well Piko PCR Plates, clear,belong to a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.

Highlights

• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 24-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 96-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions

Applications

• PCR

Related Products
Piko PCR Plate, 24-well, clear
Piko PCR Plate, 24-well, white
Plate Frame for 24-well Piko PCR Plates, white
For Research Use Only. Not for use in diagnostic procedures.
Specifications
BarcodeNo Barcode
Certifications/ComplianceCertified RNase/DNase-free
Packaging50 Plate
Product LinePiko
ColorClear
Product TypePCR Microplate
For Use With (Equipment)Piko Thermal Cycler, PikoReal Real-Time PCR System
MaterialPolypropylene
No. of Wells24
Reaction SpeedLow profile
Unit Size50 plates