Informational:Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Thermo Scientific™

Piko PCR Plate, 96-well, white

Catalog number: SPL0961
Have Questions?
Thermo Scientific™

Piko PCR Plate, 96-well, white

Catalog number: SPL0961
Have Questions?
Catalog Number
SPL0961
Unit Size
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 plates
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
SPL0961
-
SPL09615050 plates
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific 96-well Piko PCR Plates, white are a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.

Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.

Features

• Ultra Thin Wall (UTW) for fast PCR/qPCR applications
• Low profile
• Designed for use with the Piko and PikoReal 96-well thermal cyclers
• Plates can be snapped into the plate frame to create a standard 384-well plate
• Compatible with standard multi-channel pipettes and liquid-handling platforms
• Well spacing and footprint conform to industry (ANSI) dimensions

Applications

• qPCR/Real-Time PCR

Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Plate Frame for 96-well Piko PCR Plates, white
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Barcode
No Barcode
Certifications/Compliance
Certified RNase/DNase-free
Packaging
200 Plate
Product Line
Piko
Color
White
Product Type
PCR Microplate
For Use With (Equipment)
Piko Thermal Cycler, PikoReal Real-Time PCR System
Material
Polypropylene
No. of Wells
96
Reaction Speed
Standard

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text